Would you like to log in again?
Mixed Flagellates, Selected Slide
Supplier: Xinbai Shi
920110
470030-398EA 9 USD InStock
470030-398
Mixed Flagellates, Selected Slide
Slides Prepared Slides
Demonstrates Morphological Differences
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Flagella of five different Protist species represented; specially stained.
Contents:
Chlamydomonas reinhardi
Dunaliella saline
Euglena gracilis
Haematococcus pluvalis
Chilomonas paramecium
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty