Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Model Hot Air Balloons Kit
Supplier: Avantor
WARD470101-124
470101-124KT 99.99 USD InStock
470101-124
Model Hot Air Balloons Kit
Educational Classroom Kits and Activities
How does a hot air balloon stay afloat? Let your students find out for themselves as they create their own hot air balloon model using simple, everyday items. This fun and educational lab is the brainchild of California teacher, Art Morrill.
Once students complete the balloons, which are made with panels of tissue paper, they can use a stove pipe and burner to provide the heat necessary for the balloon rise. Have contests to see whose balloon stays afloat the longest.
Students practice measuring ambient (exterior) and interior air temperature, determining mass and calculation of lift. Includes detailed instructions for 10 student groups. Grades 7–12.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty