Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Ward's® Modeling Copper Extraction and Reclamation Lab Activity
Supplier: Avantor
WARD470175-690
470175-690EA 164.99 USD InStock
470175-690
Ward's® Modeling Copper Extraction and Reclamation Lab Activity
Educational Classroom Kits and Activities
Acquaint Students with Environmental Problems Created By Mining
  • Demonstrate remediation techniques for mining waste
  • Linked to a real world scenario
  • Includes a teacher's guide
During the past century, the mining industry has left behind tailing piles, which can contaminate the environment. With this kit, students perform two labs addressing the current procedures involved with removing metal wastes and restoring the affected area to its original conditions. Modeling a method used to profitably extract leftover copper from mine tailings, students learn how to conduct this procedure while limiting the environmental damage. Students then perform an experiment to determine what percentage of biosolids mixed with tailings would provide the best substrate for the growth of ryegrass, to reclaim copper tailing piles without the use of topsoil and provide habitat for wildlife. Includes a teacher’s guide, student copymasters, and materials for five setups.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty