Would you like to log in again?
Denoyer-Geppert® Multi-Torso Muscular Torso Models
Supplier: DENOYER-GEPPERT SCI CO SE
0719-F 0716-00 0712-00 0719-M 0718-00 0719-E 0719-P
470007-866EA 468.8 USD InStock
470007-866 470007-864 470007-862 470007-860 470112-592 470112-590 470007-858
Denoyer-Geppert® Multi-Torso Muscular Torso Models
Models Human Biology Models
This life-size torso encourages students to learn about how their bodies function.
  • Easy to Add More Features
  • Rotating Base
  • Six-Hundred Hand-Numbered Structures
The hand-painted, anatomically correct replica offers detailed views of the skeletal, nervous, cardiovascular, respiratory, digestive, and other systems with dissectible sections of the brain (four parts), eyeball, lung, heart (two halves), liver with gallbladder, chest muscles, stomach (two halves), intestines, kidney, vertebrae, and more. It has over 600 hand-numbered structures. Complete musculature is featured on the right side with skin, and surface features are visible on the left.
It also features quality craftsmanship, detachable breast plates, and a rotating base. The torso includes 23 detachable parts and is mounted on a hardwood, turntable base. Models may be purchased in one of three available configurations: Sexless; Male, Female, and Sexless; or Male, Female, Sexless, and Pregnant. Individual inserts are available to convert an existing model if upgrading is desired.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty