Would you like to log in again?
Ward's® Natural Recyclers: Study of Decomposition Lab Activity
Supplier: Avantor
WARD470007-674
470007-674KT 121.99 USD InStock
470007-674
Ward's® Natural Recyclers: Study of Decomposition Lab Activity
Educational Classroom Kits and Activities
Explore the Factors That Affect Decomposition
  • Determine what materials a biodegradable
  • Illustrate the factors controlling decomposition
  • Includes a teacher's guide
Students perform a series of investigations over several weeks to learn more about biodegradation of solid waste materials like paper, plastic, biodegradable plastic, natural and synthetic fibers, and more. Students study how decomposition occurs, the rate of decomposition, and the effects of different soils, temperature, and oxygen levels. The kit comes with enough materials for ten setups, a teacher’s guide, and student copymasters.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty