Would you like to log in again?
Nautilus Pompilius Shell
Supplier: TIDELINE, INC.
Ward's
532120
470027-290EA 81.35 USD InStock
470027-290
Nautilus Pompilius Shell
Specimens Preserved Specimens Preserved Animal Materials
This is the last surviving genus of an animal group with extensive fossil record.
  • Cephalopoda
  • Recent
  • Dehydrated Preserved
  • Size: 4 to 6"
  • Phillipines
The bisected Nautilus shell shows the internal chamber and siphuncle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty