Would you like to log in again?
Nostoc Slide
Supplier: Avantor
WARD470176-976
470176-976EA 6.09 USD InStock
470176-976
Nostoc Slide
Slides Prepared Slides
Cyanobacteria Genus.
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Gelatinous nodule showing filaments, within sheath.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty