Would you like to log in again?
Nylon Semimicro Test Tube Brush
Supplier: MILL-ROSE COMPANY
73200B 73200B
470016-166EA 3 USD InStock
470016-166 470120-526
Nylon Semimicro Test Tube Brush
Brushes
For test tubes 5 to 11 mm in diameter. Total length: 14.6 cm; length of fiber section: 7 cm; diameter: 1.3 cm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty