Would you like to log in again?
Ward's® Oil-Degrading Microbes Set
Supplier: VWR
WARD470179-704
470179-704KT 51.25 USD InStock
470179-704
Ward's® Oil-Degrading Microbes Set
Specimens Live Specimens Bacteria - Living Material
Study the Microbes Used in the Bioremediation of Oil Spills.
  • Useful in the process of cleaning up oil spills
This unique set features naturally occurring microbes that have an affinity for degrading petroleum products.
Penicillium sp. and Pseudomonas sp. are supplied in a convenient freeze-dried form, along with along with the media needed to rehydrate and incubate them.
The set also comes with the Working with Bacteria manual.
Ordering information: These products contain living or perishable materials.
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty