Would you like to log in again?
One Step ELISA for Diagnostics Kit
Supplier: Edvotek
267
470127-462KT 198.25 USD InStock
470127-462
One Step ELISA for Diagnostics Kit
Educational Classroom Kits and Activities
Fast and Easy ELISA Experiment
  • Fast, 1/2 the time of a typical ELISA
  • Time required: 20 minutes
Teach your students the ELISA technique in less than half the time of traditional ELISAs! This experiment eliminates the need for the primary and secondary antibody normally needed for ELISAs because the detection antibody has an enzyme linked to it directly. Simply add substrate to discover which patient is infected.
Kit includes: instructions, antigens & antibodies, substrate, phosphate buffered saline, tubes, plates, and transfer pipettes.
Required, but not included: distilled or deionized water, 37° C incubation oven, micropipettes with tips, laboratory glassware.
Packaging: Storage: Some components require refrigerator storage on arrival
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty