Would you like to log in again?
Soil Organism Study Kit
Supplier: LAB-AIDS INC (KITS) SE
33
470007-580EA 130 USD InStock
470007-580
Soil Organism Study Kit
Educational Materials Biology Educational Materials Ecology Learning Activities
What's Living In The Dirt?
  • Extract Organisms From Soil Samples
  • Identify Soil Organisms
  • Time Required: Parts Of 2 50 Minute Class Period
Students discover the teeming multitudes of living organisms that can be found in a teaspoonful of common dirt. Focusing on arthropods, they collect organisms from a soil sample, identify them, then compare different soil samples for organism population density. An exciting, open-ended activity.
Included:
Teachers Guide with MSDS
Student Worksheets and Guides
Soil Organism Extractors
Microscope slides
Collecting containers with covers, transparent
Dilute alcohol, 30 mL bottles
Materials Needed But Not Supplied: Soil Samples, Microscopes or Magnifiers
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty