Would you like to log in again?
Partial Pressure & The Ideal Gas Law Kit
Supplier: Avantor
WARD470106-814
470106-814KT 132.99 USD InStock
470106-814
Partial Pressure & The Ideal Gas Law Kit
Educational Classroom Kits and Activities
This lab uses an ingenious gas collection method to collect and determine the amount of CO2 liberated from a reaction between NaHCO3 and HCI.
The ideal gas law and Dalton’s law of partial pressure are used to determine the number of moles of CO2 gas actually produced. Includes materials for 40 student groups. Developed by Mary Gallup.
Includes: gas collecting tubes, Alka-Seltzer tables, HCI, NaHCO3, test tubes, stoppers glycerol, instructions. An electronic balance with readability to 0.01g is required but not included.
Grades 10–12.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty