Would you like to log in again?
pH Inquiry Lab
Supplier: Avantor
WARD470004-426
470004-426EA 122.09 USD InStock
470004-426
pH Inquiry Lab
Educational Classroom Kits and Activities
Experiment with pH Through Use of Indicators
  • For grades 8-10
  • Multiple experiments
In this inquiry lab, developed by teacher Mark Temmons, students explore the pH of various types of solutions using indicators. The experiments test the effect on the pH an acid as it is diluted. The final experiment explores the effect on the pH of an acid as it is neutralized.
Ordering information: Instructions included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty