Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Phagocytosis Slide
Supplier: Avantor
WARD470182-550
470182-550EA 7.75 USD InStock
470182-550
Phagocytosis Slide
Slides Prepared Slides
Illustrates Encompassing of Material
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Liver. Shows how macrophages, which are part of the reticuloendothelial system, engulf a vital dye.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty