Would you like to log in again?
Vernier® Photogate Bar Tape
Supplier: VERNIER SOFTWARE & TECH SE
TAPE-VPG
470017-524EA 16.99 USD InStock
470017-524
Vernier® Photogate Bar Tape
Adhesives
This tape allows you to broaden your study on motion.
  • Expands experiment capabilities
  • Allows you to explore motion of objects
  • Includes 3 meters of tape, four o-rings, and a guide
Use the flexible plastic tape with the Vernier Photogate to study motion while expanding your experiment capabilities. Attach the tape to objects and use it like you would a ticker tape timer or convert the bar tape into the Vernier picket fence. You can explore motions of objects like toy transportation vehicles, the motion of a person as they walk away holding the tape, and much more- the possibilities are endless. The kit includes a three meter section of bar tape that has alternating transparent and opaque bars, four O-rings to attachment, and a guide for Vernier Photogates.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty