Would you like to log in again?
Photosynthesis Microslide
Supplier: SUPERTEK SCIENTIFIC LLC
T-59 T-59-15B
470009-026PK 12.8 USD InStock
470009-026 470022-106
Photosynthesis Microslide
Slides Prepared Slides
Learn how plants convert light energy into chemical energy
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
Microslides consist of a strip of film with photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides. A detailed text folder is included. Viewers are available separately.
Ordering information: Set includes Green leaf (cs, st, 110X), Green leaf (cs, st, 350X); Root hair (wm, st, 35X); Vascular bundles (cs, st, 145X); Leaf, with and without sunlight (wm, st, 35X); Leaf, variegated (wm, 135X); and Bean seed (wm, st, 2X).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty