Would you like to log in again?
Plastic 'Pith' Ball Electroscope
Supplier: VWR International
PH0894
470111-118EA 20.2 USD InStock
470111-118
Plastic 'Pith' Ball Electroscope
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
A Cost-Effective Instrument
  • Durable
  • Quality
The plastic balls replace the more expensive pith balls and are slightly more durable but heavier. Balls are threaded and mount easily on the insulated stand. Use with friction rods or Van De Graaff generators.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty