Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Plastic Utility Cart

Supplier: LAKESIDE MANUFACTURING INC
2000
470123-546EA 645 USD InStock
470123-546
Plastic Utility Cart
Carts
Shelves are constructed of chemically-resistant polyethylene and corner posts of ABS plastic.

  • Durable and shock resistance
  • Full width handle and four swivel casters ensure easy handling and maneuverability
  • All three shelves have rims around three sides to keep contents on cart

Shelf clearance: 11". Capacity: 300 lbs.
Size: 34 7/8" x 32 1/2" x 17 1/8".
Shipping weight: 31 lbs.

Delivery information: Beige cart is shipped unassembled.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty