Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Poly ProBath Water Baths

Supplier: REVOLUTIONARY SCIENCE SE
RS-PB-300WI-FI RS-PB-100-WIFI RS-PB-100 RS-PB-300
470312-808EA 1144.3 USD InStock
470312-808 470235-878 470312-810 470006-622
Poly ProBath Water Baths
Baths Water Baths, Unstirred
The Poly ProBath is ideal for a variety of lab applications.

  • Temperature uniformity of ±0.4°C
  • Large Digital Display
  • Single two button operation
  • Temperature range: Ambient to 99.9°C
  • 2 year, repair or replacement warranty

Ideal for a variety of lab applications, this corrosion resistant polypropylene chamber is extremely simple to use and easy to clean. Whether you need to simply keep media warm or maintain precise temperature control for an experiment, this chamber will deliver consistent results. A digital microcomputer temperature controller with simple 2-button operation and 3-digit digital display is user-friendly and highly accurate. Lower water detection system sounds alarm and automatically shuts off power if water level falls beneath 1". Includes polycarbonate cover and instructions.

WiFi: The WiFi model allows logging data to the cloud or computer, and monitoring/controlling the bath remotely.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty