Would you like to log in again?
Ward's® Chemistry Polyurethane Foam Demonstration
Supplier: ALDON CORP SE
DD1020-WARDS
470222-700EA 59.09 USD InStock
470222-700
Ward's® Chemistry Polyurethane Foam Demonstration
Educational Classroom Kits and Activities
Expand Student Knowledge of Polymers
  • The perfect supplement to your chemistry lectures
  • Comes with enough chemicals to perform each demonstration five to seven times
  • Can be performed in just 10–30 minutes
  • Requires only basic labware that you provide
Ideal for illustrating foam and polymer production, as well as the use of catalysts in reactions, this dramatic demonstration will cause foam to grow 30 times the original volume of the reactants - a polyether polyol and a polyfunctional isocyanate. Use of a hood with adequate ventilation is recommended for this experiment.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty