Would you like to log in again?
Pop Beads
4336053 WARD470023-862 4336059 4336067 4336074 4336070 WARD470023-860 4336052 4336057 4336073 WARD470023-854 638-0326
470041-542PK 8.5 USD InStock
470041-542 470023-860 470041-540 470023-862 470041-552 470336-728 470041-550 470023-854 470041-548 470041-538 470041-546 470041-544
Pop Beads
Models Human Biology Models
Snap together beads.
These white pop beads with four extra holes, equally spaced in a 90° plane from the axis of attachment, are typically used to form the nucleotide backbone. Size: 10 mm dia. Package of 300.
Pop Beads
Size: 10 mm dia. Packages of 300.
Pop Bead Connectors
Simulate hydrogen bonds. Package of 100.
Magnetic Centromeres
Package of ten.
Clear Centrioles
Package of ten.
  • Multiple Colors Available
  • Connectors Available
  • Simulate Hydrogen Bonds with Special 5-Hole Beads
Build your own genetic models with these fun pop-beads. Specially designed white pop beads with four extra holes, equally spaced in a 90° plane from the axis of attachment, are typically used to form the nucleotide backbone. 10 mm diameter in size.
Pop Beads
Size: 10 mm dia. Packages of 200.
Pop Bead Connectors
Simulate hydrogen bonds. Package of 100.
Magnetic Centromeres
Package of 10.
Clear Centrioles
Package of 10.
Caution: WARNING: CHOKING HAZARD. Children under 8 years can choke or suffocate. Adult supervision is required.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty