Would you like to log in again?
Ward's® Population Dynamics Lab Activity
Supplier: Avantor
WARD470004-544
470004-544KT 133.09 USD InStock
470004-544
Ward's® Population Dynamics Lab Activity
Educational Classroom Kits and Activities
Gain an Appreciation for the Complexity of Agriculture
  • Students will grow their own radishes
  • Demonstrate important agricultural concepts
  • Includes a teacher's guide
Comprehensive Investigation for Examining Various Aspects of Plant Growth
Whatever topic you wish to cover in your plant growth studies, you’ll find an all-inclusive lab activity using Rapid Radishes to help teach the concepts. Comes with self-watering seed starter tray, Rapid Radish seeds, all necessary propagating materials, growing guide, teacher’s guide, and student copymaster.
Students will observe the effects of plant interference as they grow Rapid Radishes in different population densities and under different environmental conditions. When the plants are grown, the students will compare the leaf indexes from each treatment to determine the interference that occurred under each condition. They will then use these results to help them understand the concepts of competition, parasitism, allelopathy, and crop rotation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty