Would you like to log in again?
Potential Kinetic Track
Supplier: TONAWANDA MANUFACTURING
47615-00
470004-856EA 90.05 USD InStock
470004-856
Potential Kinetic Track
Educational Materials Physics Educational Materials Mechanics Learning Activities
Appropriate for grades 4+
This simple hands-on demonstration allows students to see the effects of height on the potential energy of a steel ball.
By releasing the ball at different heights on the raised end of the track, four different outcomes can be obtained.
Includes: metal track, 1" steel ball.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty