Would you like to log in again?
Premium Grade Culture Slides with Cavities
Supplier: United Scientific Supplies
CS3X11-PK/12
470122-938PK 13 USD InStock
470122-938
Premium Grade Culture Slides with Cavities
Slides Microscope Slides
Observe Aquatic Organisms Microscopically
  • Quality slide making supplies
  • Traditional size and material
Quality non-corrosive glass slides with ground and polished edges. All feature one or two ground and polished spherical cavities. Cavities are 15 to 18 mm in diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty