Would you like to log in again?
Promega DNA
Supplier: Promega
G1711 D1501 G1721
470191-544EA 136.09 USD InStock
470191-544 470180-828 470177-170
Promega DNA
Electrophoresis Reagents Molecular Weight Markers
Promega DNA
These high quality DNA Markers were created by digesting specified DNA to completion with one or more restriction enzymes, complete with heat inactivation. The recommended loading for each is 1μl per lane. Used for DNA electrophoresis labs. Note: Perishable. Store frozen. Shipped next day air on ice. A ship date is required.
Lambda DNA
Lambda DNA c1857 Sam7is isolated from infected e.coli strain W3350 and typically used as a molecular weight marker during DNA electrophoresis. Will yield typical patterns when cut with restriction enzymes. Uncut size is 48.5 kb. Note: Storage at -20°C required, ship date required. Available to U.S. customers only.
Lambda DNA/ HindIII Digest
This high quality DNA marker is created by digesting Lambda DNA to completion with Hind III restriction enzyme, complete with heat inactivation. Great for use as a moleduclar weight marker during DNAelectrophoresis, it is supplied with 1ml Blue/Orange 6X loading dye. Recommended loading is 1 μl per well. Note: Storage at -20°C required, ship date required. Available to U.S. customer only.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty