Would you like to log in again?
Ward's® Protist Form and Structure Set
Supplier: VWR
WARD470180-214
470328-432KT 89.99 USD InStock
470328-432 470180-214
Ward's® Protist Form and Structure Set
Specimens Live Specimens Protists - Living Material
Living Cultures Plus Comprehensive Materials for In-Depth Study.
Excellent for independent study or review, our Form and Structure Set includes not only a high-quality living culture to examine, but also a WARD’S Explano-Mount set for each culture, which includes a microscope slide of the organism for reference and a structure guide with background information. In addition, it also comes with a Form and Structure Sheet copymaster, the Working with Algae and the Working with Protozoa manuals, and materials for 35 students. Cultures included are Amoeba, Euglena, Paramecium, Spirogyra, and Volvox.
The kit is also available for purchase with a link to a NewPath digital lesson, which includes a variety of resources on the topic, including background information, illustrations and self-assessments.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty