Would you like to log in again?
PurTest™ Water Test Kits
Supplier: AMERICAN WATER SERVICE LLC
151649
470145-128CS 746.45 USD InStock
470145-128
PurTest™ Water Test Kits
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Provides Fast, Reliable Results
Test for contamination above safe EPA standards. You can test lead levels above 15 ppb and pesticide levels of 12 different Triazine elements down to 3 ppb. The Home Water Analysis tests for 11 different contaminants and includes a handbook. Each test kit comes with two tests for each factor, except for the Pesticides Kit and the lead and coliform tests in the Home Water Analysis Kit, which contain only one test.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty