Would you like to log in again?
Radiation Kit
Supplier: THE SCIENCE SOURCE SE
16226
470039-426KT 661 USD InStock
470039-426
Radiation Kit
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Used for experiments dealing with heat absorption of light surfaces versus dark surfaces
Students fill both cans with water and place a thermometer in each. They turn on light at 10 cm distance and observe how temperature rises in the black can (absorbs heat from light source) and remains essentially the same in the silver can (reflects light).
In a reverse experiment, the rates of cooling can be determined after the light is turned off.
Each kit Includes a 16 oz. black metal and a 16 oz. shiny metal can, two insulating lids, two thermometers (-20° to 50°C), and a 120V, AC clamp lamp, with reflector and 100 watt light bulb.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty