Would you like to log in again?
Ray Box
Supplier: SCIENCE INTERACTIVE GROUP
50-055
470007-096EA 63.55 USD InStock
470007-096
Ray Box
Educational Materials Physics Educational Materials Optics Learning Activities
The innovative design of this newly redesigned ray box was created to allow for a wide range of optics experiments.
The plastic body of the Ray Box houses a slide-out carrier that accommodates slits, a collimate lens, color filters, and a triple color slide. The Ray Box comes with a 10W festoon lamp, a single slit, a double slit, a triple slit, a wide slit, a triple color filter slide, ten color filters and a collimate lens. The Ray Box is 115V AC. Appropriate for Grades 5-9.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty