Would you like to log in again?
Ray Box
Supplier: STEINER ENTERPRISES SE
SE1021-00
470039-614EA 212.1 USD InStock
470039-614
Ray Box
Educational Materials Physics Educational Materials Optics Learning Activities
This updated Ray Box uses five super bright LEDs for bright, white rays that are clearly visible.
  • Metal Housing
  • Bright LEDs
  • Ideal for Introductory Ray Optic Studies
  • No Focusing Adjustment
  • Low Voltage for Safety
Unlike the monochromatic laser beams of other ray boxes, the white rays can also be used for color studies. The super bright LEDs also use less power and run much cooler than an incandescent lamp, providing a much longer life with no burned out lamps to change.
Designed for easy student use, the ray box does not require any focusing adjustment — simply plug it in and select from one, three, or five rays. It operates on safe, low voltage from a wall mount power supply for your safety.
Ordering information: This kit includes one of each: a biconvex lens, equilateral prism, rectangular block, and semicircle block.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty