Would you like to log in again?
Resonance Apparatus
Supplier: Avantor
470016-808EA 136.09 USD InStock
470016-808
Resonance Apparatus
Educational Materials Physics Educational Materials Optics Learning Activities
An exceptionally durable apparatus for determining the wavelength and velocity of sound.
  • Water reservoir may be raised or lowered to adjust the air column
  • Complete with instructions for assembly and use
An 80-cm transparent, graduated plastic tube is connected to a water reservoir. Both are securely mounted to a metal support. A tuning fork is also needed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty