Would you like to log in again?
Respiration of Yeast Lab Activity – A Student Designed Experiment
Supplier: Avantor
WARD470101-692
470101-692KT 127.99 USD InStock
470101-692
Respiration of Yeast Lab Activity – A Student Designed Experiment
Educational Classroom Kits and Activities
The study of bread ties in such diverse disciplines as biology, technology, history and cultural mores, as well as the scientific method and the importance of following proper lab procedure.
  • Developed by teacher Barbara Holbart
  • Grades 9 to 12
In this lab, students observe the expansion of bread dough as a result of yeast respiration. They then design an experiment to test the effect of one variable (temperature, light, concentration, type of yeast, additives, etc.) on that respiration. Includes materials for 40 student groups and instructions.
Caution: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty