Would you like to log in again?
Rigid Tubing
Supplier: CURBELL PLASTICS
2059900
470050-212EA 4.35 USD InStock
470050-212
Rigid Tubing
Tubing Transfer Tubing
This top quality butyrate tubing comes in 6" lengths, has an outside diameter of ⁷/₃₂" and an inside diameter of ⁵/₃₂". Smooth ends.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty