Would you like to log in again?
RS-102 Microfuge
Supplier: REVOLUTIONARY SCIENCE SE
RS-102
470005-330EA 569 USD InStock
470005-330
RS-102 Microfuge
Centrifuges Microcentrifuges
Fast Enough for DNA Research, But Inexpensive Enough for Any Lab.
  • Lightweight and compact
  • Round shape saves bench space
  • Oversized soft rubber base reduces noise and vibration
This microfuge features forced-air ventilation to ensure that specimens, rotor, and motor stay cool, even during extended runs. The easy-load rotor lets users load tubes vertically, yet spins in a 45° position. The unit slows down from maximum RPM to zero in approximately 20 seconds, and a safety switch instantly cuts power when the hinged lid is lifted. The large, clear polycarbonate cover provides complete rotor visibility. Force: 6,238 x g.
16 position rotor receives 1.5 or 2.0 ml tubes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty