Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Ward's® Chemistry Drosoph-Asleep
Supplier: VWR
AA0357-100ML
470300-994EA 32.35 USD InStock
470300-994
Ward's® Chemistry Drosoph-Asleep
Specimens Live Specimens Animals - Living Material
A Quick and Safe Method to Anesthetize Fruit Flies
Safer than ether, this solution effectively immobilizes flies without sterilizing or killing the specimens. Simple to use, a few drops of the solution placed on some blotting paper or cotton placed inside the container will put flies quickly to sleep in their culture vials. 100 mL bottle.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty