Would you like to log in again?
Ward's® Screening Hemoglobin for Sickle Cell Lab Activity
Supplier: Avantor
470024-036KT 112.89 USD InStock
470024-036
Ward's® Screening Hemoglobin for Sickle Cell Lab Activity
Educational Classroom Kits and Activities
Identify a protein defect using agarose electrophoresis.
  • Students Simulate Protein Electrophoresis
  • Materials for Eight Lab Groups
  • Time Required: ~90 Minute
In an attempt to determine the presence of the sickle-cell anemia trait, students will use agarose electrophoresis to separate several simulated hemoglobin protein samples.
They will learn to identify and differentiate between normal hemoglobin and sickle-cell hemoglobin. They will also learn to identify those who are heterozygous for the sickle-cell gene.
Accessories information: An agarose electrophoresis chamber, power supply, and 10 μL micropipets, required, are available separately.
Ordering information: Eight gels are included in the kit as well as a teacher’s guide and student copy master.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty