Would you like to log in again?
CENCO® Second Law of Motion Apparatus
Supplier: STEINER ENTERPRISES SE
0877R
470218-760PR 27.75 USD InStock
470218-760
CENCO® Second Law of Motion Apparatus
Educational Materials Physics Educational Materials Mechanics Learning Activities
This apparatus provides an exciting example of Newton's Second Law.
  • Heavy Cast Metal Base Prevents Unnecessary Movement
  • Two Spring Setting Vary the Launch Velocity
  • For Demonstrating Newton's Second Law
  • Two Steel Ball Provided
  • Instructions Provided
The unit is based on a substantial metal casting, and the entire mechanism is visible. The spring gun offers two tensions for two launch velocities.
The 25 cm long unit has a 10 mm rod.
Ordering information: This kit includes two steel balls and instructions. A support stand and clamp are required but not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty