Would you like to log in again?
Ward's® Seed Dispersal Lab Activity
Supplier: Avantor
WARD470024-222
470024-222KT 79.99 USD InStock
470024-222
Ward's® Seed Dispersal Lab Activity
Educational Classroom Kits and Activities
Students examine evolutionary adaptations for seed distribution with this kit.
  • Activity Employs NGSS Life Science DCIs
  • Students Observe How Plants Improve Offspring's Chance for Survival
  • Pre and Post-Laboratory Assessments Included
  • Enough Seed Sets for Ten Setups
  • Estimated Lab Time—Part One: 15 Minutes, Part Two: 45 Minutes
Students observe physical properties of various seeds and draw correlations between structure and dispersal strategies, such as wind, animals, and water, as they investigate the various methods plants employ to optimize the survival of offspring.
The kit includes a teacher’s guide, a student copymaster, and enough seed sets for ten setups.
Ordering information: This kit includes seed sets, clear plastic containers, simulated fur, and a tape measure. A desktop electric fan, a timer that displays seconds, and a pan, basin, or bucket are needed but not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty