Would you like to log in again?
Somso® Osteoporosis Model
Supplier: MARCUS SOMMER
QS 66/4
470030-094EA 389.75 USD InStock
470030-094
Somso® Osteoporosis Model
Models Human Biology Models
Study Physical Traits Of Prevalent Disorder
  • 5 Year Manufacturers Warranty
The possibility of developing osteoporosis is of great concern to women as they grow older. Now you can view the difference between a healthy lumbar vertebra and one affected by osteoporosis with one high-quality model. Cast from real specimens, the model is life size, and each vertebra is frontally sectioned and can be opened for an internal view. Mounted on a base, each vertebra can be removed for up-close study.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty