Would you like to log in again?
Spectacular Reactions Demonstrations
Supplier: Avantor
WARD470175-920
470175-920KT 169.99 USD InStock
470175-920
Spectacular Reactions Demonstrations
Educational Classroom Kits and Activities
Get Spectacular Reactions from Your Students When They Witness These Surprising Demonstrations
  • For grades 10-11
  • Have students take part in hands-on activities
Produce purple flames, turn a beaker ice cold, make a candle light spontaneously, and transform a “green lion” to rust and then to thick white slime all with one kit. Your students will enjoy learning the principles of displacement and redox reaction with activities guaranteed to make a lasting impression. Developed by Stewart Tick.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty