Would you like to log in again?
Plastic Cuvettes (Visible Range)
Supplier: VERNIER SOFTWARE & TECH SE
CUV
470120-596PK 23.99 USD InStock
470120-596
Plastic Cuvettes (Visible Range)
Cuvettes Spectrophotometer Cuvettes
A package of 100-3.5 ml cuvettes (with 20 lids).
For use with our colorimeter, SpectroVis Plus, and vernier spectrometer.
For educational use only.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty