Would you like to log in again?
Spectrum Analysis Chart
Supplier: SCIENCE INTERACTIVE GROUP
SP-187
470005-828EA 50 USD InStock
470005-828
Spectrum Analysis Chart
Charts
Details the Spectra of Ten Elements
  • Heavy grade paper
Study the electromagnetic, visible, and solar spectrums, the Doppler effect, the absorption spectra of solutions, and the bright-line spectra of ten elements. The drawings illustrate methods of producing various spectra and practical applications of spectrum analysis. The scale under each spectrum runs from 3,900–7,600 Å. The color chart, constructed of heavy paper, is bound at the top and bottom with loops for hanging.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty