Would you like to log in again?
Spectrum Tube Carousel
Supplier: STEINER ENTERPRISES SE
SE1029-00
470013-434EA 416.05 USD InStock
470013-434
Spectrum Tube Carousel
Educational Materials Physics Educational Materials Optics Learning Activities
This carousel is a unique device for storing and using spectrum tubes.
  • Safe Design
  • Carousel Holds up to Eight Tubes
  • On-Board Power Supply
This carousel holds eight spectrum tubes and activates one tube at a time when connected to the on-board power supply.
To view the other tubes, simply rotate the carousel, so a different tube can be activated. This allows for quick and easy holding and changing tubes for an experiment.
Ordering information: Spectrum tubes sold separately. The Spectrum Tube Carousel does not come supplied with any Spectrum tubes.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty