Would you like to log in again?
Spinal Cord with Ganglion Slide
Supplier: Avantor
WARD470189-622
470189-622EA 7.45 USD InStock
470189-622
Spinal Cord with Ganglion Slide
Slides Prepared Slides
Illustrates components of nerves and nerve tissue
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Shows dorsal and ventral nerve roots and ganglion. Stained for clear distinction of cellular elements, nerve fibers, and connective tissue.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty