Would you like to log in again?
Spinal Cord, Four Levels Slide
Supplier: Avantor
WARD470182-714
470182-714EA 19.75 USD InStock
470182-714
Spinal Cord, Four Levels Slide
Slides Prepared Slides
Compare and contrast nervous tissue
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. The slide combines cross sections of cervical, thoracic, lumbar, and sacral regions. It is stained for myelin sheaths of fibers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty