Would you like to log in again?
Spinal Ganglion Slide
Supplier: Avantor
WARD470177-604
470177-604EA 10.65 USD InStock
470177-604
Spinal Ganglion Slide
Slides Prepared Slides
Nervous System Structure.
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Typical slide for large nerve cells with capsules and myelinated nerve fibers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty