Would you like to log in again?
St. Louis Demo Motor
Supplier: VWR International
PH1236
470039-450EA 77.85 USD InStock
470039-450
St. Louis Demo Motor
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Students use the St. Louis Demo Motor to research fundamental principles of electric motors and generators.
  • Motor Demonstrates Effects of Pole Separation
  • Two-Pole D.C. Armature
  • Brush Holder Can Be Rotated for Experiments
  • Two Heavy Binding Post Terminals
  • Permanent Bar Magnets
This motor consists of a two-pole D.C. armature. The brush holder, with two heavy binding post terminals, can be rotated for experiments.
Permanent bar magnets are fixed in the horizontal plane to demonstrate effects of pole separation. A power supply is required but not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty