Would you like to log in again?
Stigeoclonium
Supplier: VWR
470179-808EA 9.5 USD InStock
470179-808
Stigeoclonium
Specimens Live Specimens Protists - Living Material
Freshwater. Filamentous. Produces zoospores. Basic culture solution.
Characterized by green plastids and cell walls composed of cellulose.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty