Would you like to log in again?
Ward's® Stormwater Floodplain Simulation System
Supplier: Avantor
8551-5812 8551-6136 8551-5813
470164-072KT 1715.79 USD InStock
470164-072 470165-328 470165-332 470165-330
Ward's® Stormwater Floodplain Simulation System
Educational Classroom Kits and Activities
This system helps students understand the critical role that floodplains play in the life of a watershed and the impact of unplanned development and human activity in key areas through innovative hands-on simulations.
  • Realistic Recreation with Student-Friendly Features
  • Kit Features Eight Activities
  • Detailed User's Guide Included
  • Large Acrylic Tank Included
  • Students Work Together to Find Solutions to Floodplain Issues
Developed for education and outreach in cooperation with the Michigan Stormwater-Floodplain Association, this large-scale model allows students to create, test, and visualize how various factors within the watershed can impact storm water runoff and flooding. Through the use of the model, students explore the value of retention ponds and wetlands in flood management. They construct their own levees and witness how stream flow is affected during times of high runoff. They work as a team to find and test solutions to a variety of floodplain problems simulated by the model. A large, clear acrylic tank features a durable, hand-painted resin landform insert and two different rainmaker trays to create different rainfall intensities.
Three custom acrylic “plug and play” headwater trays for wetland, parking lot, and retention-pond simulations also come with a variety of accessories and models for enacting realistic environmental scenarios. Additional materials are provided for constructing levees and other landscape settings. A custom riser allows for stepped changed in slope and broader experimentation, and a staff gage is mounted for clearly and easily measuring river levels.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty