Would you like to log in again?
String Vibrator, AC
Supplier: THE SCIENCE SOURCE SE
15250
470006-552EA 124.65 USD InStock
470006-552
String Vibrator, AC
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
This device will drive one end of one or more taut strings at a steady 60 Hz, allowing the formation and investigation of standing waves. The novelty offered by this 60 Hz vibrator is that several hanks of fine line can be attached at the same time so that students can see how the standing waves formed are controlled by the length and tension in the line. Five hanks of braided line are included for good qualitative and quantitative data.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty